Learn More
Description
The Ha4/8 antibody is specific for the keyhole limpet hemocyanin (KLH).
Host Species: Hamster
Clone: Ha4/8
Isotype: IgG2, λ 1
Immunogen: Keyhole limpet hemocyanin
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | Ha4/8 |
| Concentration | 0.2mg/ml |
| Conjugate | PE-CF594 |
| Host Species | Hamster |
| Immunogen | Keyhole limpet hemocyanin (KLH) |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Content And Storage | 4°C. Store in dark. |
| Isotype | IgG2 λ1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.