Learn More
American Research Products Inc Hamster Interleukin 6 IL 6 EL
Supplier:  American Research Products Inc CSBE14304HA
Encompass
Hamster Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26735