Learn More
MyBioSource PL-A2, Hamster ELISA Kits
SDP
Supplier:  MyBioSource MBS028224
Encompass_Preferred
The MyBioSource Hamster Phospholipase A2 ELISA Kit quantifies Phospholipase A2 in various biological fluids using a specific antibody-coated 96-well plate. After binding and washing steps, a colorimetric reaction proportional to the Phospholipase A2 levels is measured at 450 nm.
Catalog No. NC2831179
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.