Learn More
MyBioSource Hamster ELISA Kit for Salicylic Acid (SA)
SDP
Supplier:  MyBioSource MBS075683
Encompass_Preferred
The MyBioSource Hamster Salicylic Acid ELISA Kit quantitatively measures Salicylic Acid in various hamster biological fluids using a competitive assay with a 96-well plate. It features a detection range of 0.156-10 ng/mL and a sensitivity of 0.094 ng/mL. The colorimetric readout at 450nm reflects Salicylic Acid concentration.
Catalog No. NC2831178