Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Hand Applicator

Catalog No. 19072012 Shop All 3M Products
Change view
Click to view available options
Color:
Blue
Gold
Description:
Firm edge
Soft Edge Hand Applicator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description
19-072-012 Blue Soft Edge Hand Applicator
19-072-096 Gold Firm edge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-072-012 Supplier 3M™ Supplier No. 7100038651
Add to Cart
Add to Cart
This item is not returnable. View return policy

3M™ Hand Applicator helps to hand apply premasking tape or conform graphic film over very lighted textured substrates

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Description Soft Edge Hand Applicator
For Use With (Application) Hand applying premasking tape; Conforming graphic film over very lighted textured substrates such as FRP
Model PA1-B
Quantity 25/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.