Learn More
United Scientific™ Hand Centrifuge
SDPSCI_ED
Catalog No. S99443 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity:
4 Tubes
Catalog No. Capacity
S99443 4 Tubes
1 options
Catalog No. S99443 Supplier United Scientific™ Supplier No. CENTFG1
Add to Cart
Add to Cart
Hand-driven Centrifuge comes with a table-mounting clamp.
Hand-driven centrifuge comes with a table-mounting clamp.
  • Features a painted metal body and metal head.
  • Comes complete with four aluminum shields.
  • Easily disassembles for storage in specially designed foam box.
  • Crank is geared to head at 16:1 ratio.
  • A crank rate of 12 turns in 10 seconds yields a head speed of 1152 RPM.
Specifications
Capacity 4 Tubes
Max. RCF Manual Crank
Max. Speed 1,152 rpm
Refrigerated No
Model CENTFG1
Product Type Hand Centrifuge