Learn More
Eisco™ Hand-Driven Van de Graaff Generator and Accessories
SCI_ED
Catalog No. S27749 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Spare Belt
Spare O-ring Belt
Van de Graaff Generator
Van de Graaff Kit, Winter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S27749 Spare Belt
S27752 Spare O-ring Belt
S95974 Van de Graaff Generator
S27893 Van de Graaff Kit, Winter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S27749 Supplier Eisco™ Supplier No. PH0918BL
Only null left
Add to Cart
Add to Cart
For Demonstrating Electrostatic Principles
  • Apparatus consists of 7 in. (17.5 cm) metal sphere, hand-operated pulley and insulating plastic support column on base
  • As generator is operated, charge builds up on metal sphere
  • With sufficient charge, electrical breakdown of air surrounding sphere occurs
  • Various replacement belts are available, sold separately
Specifications
Description Thickness: 1.5 mm
For Use With (Application) Demonstrating Electrostatic Principles
For Use With (Equipment) Van De Graaff Hand Crank Generator (S95974)
Product Type Spare Belt
Width (English) 2.45 in.