Learn More
United Scientific Supplies Hand Held Generator
SDPSCI_ED
Catalog No. S23977 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
6V DC
Catalog No.
S23977 6V DC
1 options
This item is not returnable. View return policy
Catalog No. S23977 Supplier United Scientific Supplies Supplier No. GENH02
Add to Cart
Add to Cart
This item is not returnable. View return policy
6V DC
This 6V DC hand held generator can deliver up to 400mA current. The generator is equipped with a polarity coded plug-in lead with alligator clips. The clear plastic housing allows students to watch the coils, terminals, and gears. Students get a hands-on demonstration of the Second Law of Thermodynamics. Includes activity guide.
Specifications
Product Type Generator, Handheld