Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ HAND MEDIC™ Professional Skin Conditioner
SDP

Catalog No. 19043856 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
147.87 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-856 147.87 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-856 Supplier GOJO™ Supplier No. 815012
  • $16.66
    $13.65 / Each

    Save $3.01

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $173.90
    $142.65 / Case of 12 EA

    Save $31.25

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prevent dry, cracked skin by maintaining skin moisture levels. Professional skin conditioner is specially formulated for industrial environments.

  • Fragrance-free skin conditioner
  • Specially formulated for industrial environments
  • Helps prevent dry, cracked skin by maintaining skin's moisture level
  • Absorbs quickly with no greasy after-feel
  • Silicone and fragrance free
  • Dermatologist tested
  • Latex glove compatible
  • NSF E4 Registered
TRUSTED_SUSTAINABILITY

Specifications

Product Type Skin Conditioner
Color White
Form Lotion
For Use With (Application) Helps prevent dry, cracked skin by maintaining skin moisture level
Quantity 12/Pk.
Size 5 oz.
Volume (Metric) 147.87 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.