Learn More
United Scientific™ Hand Microtome
SDPSCI_ED
Catalog No. S17448B Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Type:
Hand Microtome
Catalog No. Type
S17448B Hand Microtome
1 options
Catalog No. S17448B Supplier United Scientific™ Supplier No. MCT001
Add to Cart
Add to Cart
The Hand Microtome allows you to cut sections to 10 microns.
  • Easy to use, this microtome lets you cut sections to 10 microns
  • Equipped with a clamping mechanism for securing various shapes and sizes of objects
  • Supplied in a cardboard storage box and includes a tempered steel razor knife
  • Includes instructions for use
Specifications
Type Hand Microtome
Includes Instructions