Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hand Rule Demonstrator
SDPSCI_ED

Catalog No. S15866 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15866 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15866 Supplier United Scientific™ Supplier No. HANDEM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Engaging visual demonstration of a magnet and an induced current.

  • Vividly demonstrate the interaction of a magnet and an induced current. As electrons flow in a wire, they create a magnetic field. When this current-carrying wire is near a permanent magnet it causes magnetic repulsion and motion is created.
  • The apparatus works on 3 V to 6 V DC and has a switch to cause a temporary current in the magnetic coil
  • Includes activity guide
  • Note: power supply and galvanometer not included
  • Size assembled: 7.5 x 5.5 x 11 in.

Specifications

Material Plastic
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Hand Rule Demonstrator

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.