Learn More
Description
- Steel box keeps your sanitizer protected in place
- Lockable design prevents people from taking the hand sanitizer
- Box includes mounting hardware to keep it in place
- Hand sanitizer not included
Specifications
Specifications
| Product Type | Lockable Hand Sanitizer Box |
| Color | Blue on White |
| Dimensions (D x W x H) Interior | 5.25 x 3.5 x 5.25 in. |
| For Use With (Application) | Hand Sanitizer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.