Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Hand Skeleton Model
SCI_ED

Catalog No. S06697 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
4.5 x 4.5 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S06697 4.5 x 4.5 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06697 Supplier Walter Products™ Supplier No. B10210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fully articulated with wire.

  • Mounted securely on a base
  • Shows the distal portions of the ulna and radius
  • Slightly flexible to portray the natural movement of the human hand
  • Dimensions: 4.5 x 4.5 x 10 in. (11.4 x 11.4 x 25.4cm)

Specifications

Dimensions (L x W x H) 4.5 x 4.5 x 10 in.
Includes Distal portions of the ulna and radius
Product Type Model, Skeleton, Hand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.