Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597267 Supplier Invitrogen™ Supplier No. PA597267
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHPAHPMLHE PFLFGPASRC HQERPYFQSW LLSPADAAPD FPAGGPPPAA AAAATAYGPD ARPGQSPGRL EALGGRLGRR KGSGPKKERR RTESINSAFA ELRECIPNVP ADTKLSKIKT LRLATSYIAY LMDVLAKDAQ SGDPEAFKAE LKKADGGRES KRKRELQQHE GFPPALGPVE KRIKGRTGWP QQVWALELNQ; Positive Samples: SH-SY5Y, A-549, NIH/3T3; Cellular Location: Nucleus, nucleolus, nucleoplasm.

HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAND1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HAND1
Gene Accession No. O96004, Q64279
Gene Alias bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Ehand1; extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; HAND1; heart and neural crest derivatives expressed 1; heart and neural crest derivatives expressed transcript 1; heart- and neural crest derivatives-expressed protein 1; Helix-loop-helix transcription factor expressed in extraembryonic mesoderm and trophoblast; Hxt; Th1; Thing1; thing-1
Gene Symbols HAND1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 16-215 of human HAND1 (NP_0048121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15110, 9421
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.