Learn More
TSI Incorporated AeroTrak Handheld Particle Counter
Supplier:  TSI Incorporated 930603
Encompass
The TSI AeroTrak Handheld Particle Counter Model 9306 offers the most features and flexibility for customers interested in versatile handheld particle contamination monitoring. The Model 9306 features an ergonomic handle with thumb controls, for easy one-hand operation. The 3.6-inch (9.1-cm) color touch screen interface makes it easy to configure and operate. The Model 9306 can generate Pass/Fail reports for ISO 14644-1, EU GMP Annex 1, and FS209E. Particle count data can be conveniently viewed on screen, downloaded using TrakPro Lite Secure Software, or printed directly to an optional external printer.
Catalog No. NC1848666
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.