Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HandPRO™ Sable600™ Black Nitrile Gloves
GREENER_CHOICE

Catalog No. 17010398 Shop All Hourglass International Inc Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-398 Medium
17-010-396 X-Small
17-010-397 Small
17-010-399 Large
17-010-400 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-010-398 Supplier Hourglass International™ Supplier No. 602
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nitrile exam gloves that are created from accelerator-free LOW DERMA™ technology nitrile, deliver improved stretch, chemical resistance, and tactile sensitivity. Bold black color hides stains during use and makes light-colored contaminants more visible.

  • Made from patented LOW DERMA technology nitrile to reduce your risk of glove-related allergic contact dermatitis
  • Accelerator-free nitrile with no thiurams, carbamates or MBTs
  • Chemotherapy drug permeation tested
  • Fentanyl permeation tested (>240 minutes; 510(k) Granted by U.S. FDA)
  • Excellent chemical resistance
  • High tactile sensitivity
  • Finger-textured for a secure grip
  • Bold black color hides stains during use and makes light colored contaminants more visible
  • Meets ASTM D6978 and USP 800 PPE requirement for use of chemotherapy gloves when handling hazardous drugs
  • Meets FDA and EU requirements for food handling
  • 300-count box maximizes space, reduces waste and minimizes cost
  • Fits standard size glove box holder
  • Powder-free

Specifications

Certifications/Compliance ASTM D6319 Nitrile Exam Gloves for Medical Application; ASTM D6978 Chemotherapy Drugs and Fentanyl Citrate Tested; ASTM F1671 Viral Penetration Test
Certifications/Compliance (Europe) EN 455 Nitrile Exam Gloves for Medical Application
Chemotherapeutic Rated Yes
Chlorinated Yes
Accelerator Free Yes
Applications Healthcare, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3.1 to 3.9 mil
Finger Thickness (Metric) 0.08 to 0.10 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.3 to 2.7 mil
Palm Thickness (Metric) 0.06 to 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (English) 3.1 to 3.9 mil
Thickness (Metric) 0.08 to 0.10 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Tensile Strength 30 to 34 Mpa
Length (English) 9.5 in.
Length (Metric) 24.5 cm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.