Learn More
Grainger Balaclava Black Fleece
Supplier: Grainger 16W660
The N-FERNO Balaclava by Ergodyne is a cold weather face mask made of 92% polyester and 8% spandex. It features a seamless single layer construction to provide warmth and wind protection without restricting vision, hearing, or breathing. The balaclava has a skull cap design that pulls over the head and covers the neck, face, and head while leaving the eyes, nose, and mouth areas open. It can be worn under a hard hat or helmet. The balaclava comes in black and is machine washable. It provides warmth and protection from wind chill down to -20°F.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.