Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Hanging Human Skeleton with Nerve Endings, Half Size
SCI_ED

Catalog No. S15014 Shop All Eisco Products
Change view
Click to view available options
Skeleton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15014 Skeleton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15014 Supplier Eisco™ Supplier No. AMCHA105H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Half Size Hanging Human Skeleton with nerve endings exhibits realistic details to make it an excellent tool for educational study, as well as for patient demonstrations.

  • Quality hardware allows for easy movement of major joints
  • Mandible is articulated and removable for closer study of jaw, teeth, and internal structures
  • Nerve endings are visible for advanced nervous system investigation
  • Arms and legs can be removed for storage and study
  • Features realistic details, textures and bony landmarks
  • Half sized human skeleton stands 34 in. tall
  • Mounted on a sturdy hanging stand
  • Made of durable and sturdy plastisol polymer
  • Skeleton is washable and easy to clean to hold up to years of handling
  • Includes key card for identifying and studying the various bones that make up a full skeleton

Specifications

Color White
For Use With (Application) Educational Study, Patient Demonstrations
Height (English) 34 in.
Height (Metric) 86 cm
Material Plastisol Polymer
Mounting Hanging Stand
Product Type Model, Skeleton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.