Learn More
Invitrogen™ Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric Recombinant Human Monoclonal Antibody (B1-8)
Shop All Thermo Scientific Products

Description
Specificity: Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).
Specifications
Specifications
| Antigen | Hapten 4-hydroxy-3-nitrophenyl acetyl NP Chimeric |
| Applications | Control |
| Classification | Recombinant Monoclonal |
| Clone | B1-8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Host Species | Human |
| Immunogen | hapten-protein conjugated NP12-chicken gammaglobulin (CG). |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.