Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Haptoglobin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524174 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524174 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524174 Supplier Invitrogen™ Supplier No. PA524174
  • $620.00
    $498.65 / Each of 1

    Save $121.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Haptoglobin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Haptoglobin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HP
Gene Accession No. P00738, Q61646
Gene Alias alpha polypeptide; Ba1-647; beta polypeptide; binding peptide; Binding peptide (BP); BP; haptoglobin; haptoglobin alpha 1S; Haptoglobin alpha chain; Haptoglobin alpha chain / alpha(1S) beta; haptoglobin alpha(1S)-beta; Haptoglobin alpha(2FS) beta; haptoglobin alpha(2FS)-beta; Haptoglobin beta chain; haptoglobin, alpha polypeptide; haptoglobin, beta polypeptide; haptoglobin; haptoglobin-like; haptoglobin-like; haptoglobin-related protein; Hp; HP-1; HP2 ALPHA2; HP2ALPHA2; HP2-ALPHA-2; HPA1S; HPR; HPT; Liver regeneration-related protein LRRG173; LOC100067869; LOC479668; MGC111141; MGC133752; preHP2; zonulin
Gene Symbols HP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 296-322 amino acids from the central region of human HP.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15439, 3240
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.