Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HARBI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558490 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558490 Supplier Invitrogen™ Supplier No. PA558490
  • $617.00
    $498.65 / Each of 1

    Save $118.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: VIEKTFRTLC SRFRCLDGSK GALQYSPEKS SHIILACCVL HNISLEHGMD VWSSPMTGPM EQPPEEEYEH MESLDLEADR IRQELM Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 91%.

Transposase-derived protein that may have nuclease activity (Potential). Does not have transposase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HARBI1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HARBI1
Gene Accession No. Q96MB7
Gene Alias C11orf77; D230010M03Rik; Harbi1; harbinger transposase derived 1; Harbinger transposase-derived nuclease; Putative nuclease HARBI1
Gene Symbols HARBI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HARBI1. Recombinant protein control fragment (Product #RP-97520).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283254
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.