Learn More
Description
- Soft lightweight material
- Hi-vis material for greater visibility
- Fits on the outside of cap style and full brim hard hats
Specifications
Specifications
| Product Type | Hard Hat Shade |
| Color | Orange |
| For Use With (Equipment) | Hard Hat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.