Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ SL Series Hard Hats, 4-point Ratchet

Catalog No. 19161137 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-161-137 White
19-161-141 Blue
19-161-142 Green
19-161-140 Yellow
19-161-138 Red
19-161-143 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-161-137 Supplier Pyramex™ Supplier No. HP14110
Add to Cart
Add to Cart

Hard Hats exceed ANSI Z89.1, Type 1, Class C, G, and E for industrial head protection; CSA Z94.1-15.

  • Low profile design
  • Tested and marked for reverse donning
  • Shell constructed from High Density Polyethylene materials
  • Rain trough on sides and back of helmet channel moisture away
  • Soft brow pad is replaceable. Replaceable suspensions and headbands also available
  • Snap Lock suspension features 'tuck away' adjustments for all day, hassle free wear
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
  • Universal accessory slots fit cap mounted ear muffs and accessories. Hats also accommodate chin straps
  • Testing: Impact, Penetration, Flammability, Electrical Insulation, Marking & Humidity

Specifications

Certifications/Compliance ANSI Z89.1
Color White
For Use With (Equipment) Earmuffs, Faceshield
For Use With (Application) Construction
Material HDPE
Product Type Cap-style Hard Hat
Size 6.5 to 8
Ventilated Not Vented
Head Style Short Brim (Cap Style)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.