Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Hardwood Meter Scale
SCI_ED

Catalog No. S24856 Shop All Eisco Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S24856 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24856 Supplier Eisco™ Supplier No. PH0064E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hardwood Meter Scale with graduations on both edges in centimeters and millimeters.

  • Horizontal reading with brass end
  • Made of hardwood

Specifications

Grade K to Undergraduate
Includes Graduations in centimeters and millimeters on both sides, brass end
Material Hardwood
Length (English) 3.3 ft.
Length (Metric) 1 m
Class Size 1 Lab Group
Product Type Meter Scale
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.