Learn More
Hardy Diagnostics™ HardyDisks™ Cefiderocol, FDC30, Fetroja
HardyDisk AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
Supplier: Hardy Diagnostics™ Z9435
This item is not returnable.
View return policy
Description
- HardyDisk™ AST Disks are used for semi-quantitative in vitro susceptibility testing by the agar diffusion test procedure (KirbyBauer) of rapidly growing and certain fastidious bacterial pathogens
- HardyDisk™ AST Disks are prepared by impregnating high-quality 6mm diameter white filter paper disks with accurately determined amounts of antimicrobics or other chemotherapeutic agents
- Disks are clearly marked on both sides with letters and numbers designating the agent and the drug content
- Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing
- Use of HardyDisk™ AST Cefiderocol 30μg (FDC30) for in vitro agar diffusion susceptibility testing is indicated when there is the need to determine the susceptibility of bacteria to Cefiderocol
- HardyDisk™ AST Cefiderocol at concentration 30μg can be used to determine the zone diameter (mm) of Cefiderocol against the following bacteria for which Cefiderocol has been shown to be active both clinically and in vitro:
- Escherichia coli
- Enterobacter cloacae complex
- Klebsiella pneumoniae
- Proteus mirabilis
- Pseudomonas aeruginosa
Specifications
Antimicrobial Susceptibility Discs | |
Antibiotic Susceptibility Sensitivity | |
50 | |
50 Pk. |