Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmaline 98.0+%, TCI America™
SDP

Catalog No. H12371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12371G 1 g
H12375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12371G Supplier TCI America Supplier No. H12371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 304-21-2
Molecular Formula C13H14N2O
Molecular Weight (g/mol) 214.268
MDL Number MFCD00004955
InChI Key QJOZJXNKVMFAET-UHFFFAOYSA-N
Synonym harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline
PubChem CID 5280951
IUPAC Name 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole
SMILES CC1=C2C(=C3C=CC(=CC3=N2)OC)CCN1

Specifications

Melting Point 251°C
Color Yellow
Quantity 1 g
Formula Weight 214.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Harmaline
RTECSNumber UU9800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.