Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmine, 98%

Catalog No. AC302970050
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC302970050 5 g
AC302972500 250 mg
AC302970010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC302970050 Supplier Thermo Scientific Chemicals Supplier No. 302970050
Add to Cart

CAS: 442-51-3 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym 7-methoxy-1-methyl-9h-beta-carboline, leucoharmine, banisterine, banisterin, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, telepathine, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, yageine, harmine, yajeine
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Melting Point 262.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 4649
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 212.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Harmine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:

Harmful if swallowed.

Harmful if inhaled.

Harmful in contact with skin.
Precautionary Statement GHS P Statement:

Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN: Wash with plenty of soap and water.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-131-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.