Learn More
Selleck Chemical LLC Harmine hydrochloride 25mg 343-27-1 Telepathine hydrochloride
SDP
Supplier:  Selleck Chemical LLC S381725MG
Encompass_Preferred
Harmine (Telepathine), a fluorescent harmala alkaloid belonging to the beta-carboline family of compounds, is a highly cell-permeant and competitive inhibitor of ATP binding to the kinase pocket of DYRK1A, with about 60-fold higher IC50 value for DYRK2. Harmine also inhibits monoamine oxidases (MAOs) and cdc-like kinases (CLKs). Harmine inhibits 5-HT2A serotonin receptor with Ki of 397 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8247
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.