Learn More
Lkt Laboratories CAS 26833-85-2, ≥98%, Cephalotaxine alkaloid originally found in Cephalotaxus; ribosomal protein synthesis inhibitor.
SDP
Supplier:  Lkt Laboratories H0169
Encompass_Preferred
Harringtonine is a cephalotaxine alkaloid originally found in Cephalotaxus hainanensis Li; it exhibits antiviral and anticancer activities. Harringtonine immobilizes initiated ribosomes, inhibiting protein translation by binding the A site of the 60 S ribosomal subunit and preventing aminoacyl-tRNA binding. Harringtonine inhibits chikungunya virus infection, suppressing replication, RNA production, and viral protein expression. Alone, harringtonine downregulates production of Mcl-1, increases cleavage of PARP, and inhibits cell growth in acute promyelocytic leukemia (APL) cells. Additionally, this compound improves the efficacy of co-administered chemotherapeutic compounds, further inhibiting proliferation and inducing apoptosis in leukemia cells.
Catalog No. NC2539830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.