Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Barricade Tapes

Catalog No. 19019951
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Red
Yellow
Dimensions (L x W):
1000 ft. x 3 in.
300 ft. x 3 in.
Displayed Text:
Caution Caution Caution
Caution Construction Area
Caution Do Not Enter
Danger Danger
Danger Do Not Enter
Danger Hazardous Area Keep Out
Danger Hazardous Material Do Not Enter
Danger Keep Out
Fire Line Do Not Cross
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W) Displayed Text
19-019-951 Yellow 1000 ft. x 3 in. Caution Do Not Enter
19-019973 Red 1000 ft. x 3 in. Danger Do Not Enter
19-068-000 Yellow 300 ft. x 3 in. Caution Do Not Enter
19-811-825 Yellow 1000 ft. x 3 in. Fire Line Do Not Cross
19-019972 Red 1000 ft. x 3 in. Danger Danger
19-019974 Red 1000 ft. x 3 in. Danger Hazardous Area Keep Out
19-019975 Red 1000 ft. x 3 in. Danger Hazardous Material Do Not Enter
19-019976 Red 1000 ft. x 3 in. Danger Keep Out
19-019-948 Yellow 1000 ft. x 3 in. Caution Caution Caution
19-019950 Yellow 1000 ft. x 3 in. Caution Construction Area
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-019-951 Supplier Harris Industries Supplier No. BT8

Please call customer service for current pricing. 1-800-766-7000

This item is not returnable. View return policy

Increase visibility of hazards to help prevent accidents

Strong, lightweight polyethylene tapes with continuous repetition of message.

  • Can be tied, stapled, or nailed into place
  • Economical and reusable
  • Caution tapes have yellow background
  • Danger tapes have red background

Order Info

Roll size: 3 in. W x 1000 ft.L (7.6cm x 304.8m). Thickness: 4mil.

Compliance

Meet OSHA 1910.144 for marking physical hazards.

Specifications

Dimensions (L x W) 1000 ft. x 3 in.
Width (English) 3 in.
Width (Metric) 7.62 cm
Thickness 4 mil
Displayed Text Caution Do Not Enter
Type Barricade Tape
Certifications/Compliance OSHA 1910.144
For Use With (Application) Increase visibility of hazards to help prevent accidents
Length (English) 1000 ft.
Length (Metric) 304.8 m
Adhesive Type Non-adhesive
Color Yellow
Material HDPE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.