Learn More
Harris Industries Barricade Tapes
Description
Strong, lightweight polyethylene tapes with continuous repetition of message.
- Can be tied, stapled, or nailed into place
- Economical and reusable
- Caution tapes have yellow background
- Danger tapes have red background
Order Info
Roll size: 3 in. W x 1000 ft.L (7.6cm x 304.8m). Thickness: 4mil.
Compliance
Meet OSHA 1910.144 for marking physical hazards.
Specifications
Specifications
| Dimensions (L x W) | 1000 ft. x 3 in. |
| Width (English) | 3 in. |
| Width (Metric) | 7.62 cm |
| Thickness | 4 mil |
| Length (Metric) | 304.8 m |
| For Use With (Application) | Increase visibility of hazards to help prevent accidents |
| Certifications/Compliance | OSHA 1910.144 |
| Displayed Text | Danger Hazardous Area Keep Out |
| Length (English) | 1000 ft. |
| Color | Red |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.