Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harris Industries Colored Delineator Tape

Catalog No. 19086347
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Black, Yellow
Red, White
Dimensions (L x W):
18 yd. x 3 in.
36 yd. x 2 in.
36 yd. x 3 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
19-086347 Black, Yellow 18 yd. x 3 in.
18-785 Red, White 36 yd. x 2 in.
19-020002 Black, Yellow 36 yd. x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-086347 Supplier Harris Industries Supplier No. HT202BY
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.

Features:

  • Alternating color stripes
  • Highly visible
  • Pressure sensitive
  • Rubber-based permanent adhesive
  • Self-wound
  • Clear laminate resists oil, dirt, chemicals and abrasion
  • 6.0mil/ 0.150mm thickness

Benefits:

  • Laminated PVC will not fade or scuff
  • Holds up to traffic and wear longer than paint
  • Can be applied quickly and easily with minimal surface preparation
  • Maintenance-free; brilliance is restored with wet mopping/washing

Red/White: Fire Protection Equipment; Black/Yellow: Physical Hazard; Black/White: Housekeeping Aisle Marking

Compliance

OSHA 1910.144

Specifications

Dimensions (L x W) 18 yd. x 3 in.
Adhesive Type Rubber-based Adhesive
Color Black, Yellow
Width (English) 3 in.
Certifications/Compliance OSHA 1910.144
Thickness 6 mil
Width (Metric) 7.62 cm
Tensile Strength 23N/cm
Temperature Range (English) 14°F to 176°F
Length (Metric) 16.4 m
Temperature Range (Metric) -10°C to +80°C
Height (Metric) 7.62 cm (3 in.)
Length (English) 18 yd.
For Use With (Application) Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
Material PVC
Type Hazard Warning Tape
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.