Learn More
Sigma Aldrich Fine Chemicals Biosciences Hartmans fxatve hstolog500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H0290500ML
Encompass
Hartman fixative which is also known as Davidson fixative contains acetic acid alcohol and formalin. It is widely used for the preservation of eye bone marrow breast testis and other tissues for histologic examination.
Catalog No. 50-290-0693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.