Learn More
Invitrogen™ HAS1 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SHG-44 whole cell, human THP-1 whole cell, rat brain tissue, rat smooth muscle tissue, rat ovary tissue, mouse brain tissue, mouse smooth muscle tissue, mouse ovary tissue, mouse small intestine tissue, mouse Neuro-2a whole cell. IHC: human lung cancer tissue, human mammary cancer tissue, human mammary cancer tissue, mouse spleen tissue, rat small intestine tissue. ICC/IF: U20S cell, U20S cell . Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | HAS1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | Has1 |
| Gene Accession No. | Q61647, Q92839 |
| Gene Alias | HA synthase 1; HAS; Has1; huHAS1; Hyaluronan synthase 1; hyaluronan synthase1; hyaluronate synthase 1; Hyaluronic acid synthase 1 |
| Gene Symbols | Has1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.