Learn More
Enzo Life Sciences HAT activity assay kit (100 tests)
SDP
Supplier:  Enzo Life Sciences ALX850326KI01
Encompass_Preferred
Histone acetyltransferases (HATs) have been implicated to play a crucial role in various cellular functions, such as gene transcription, differentiation, and proliferation. HAT Activity Colorimetric Assay Kit offers a convenient, nonradioactive system for a rapid and sensitive detection of HAT activity in mammalian samples. The kit utilizes active recombinant HAT as a positive control and acetyl-CoA as a cofactor. Acetylation of peptide substrate by active HAT releases the free form of CoA which then serves as an essential coenzyme for producing NADH. NADH can easily be detected spectrophotometrically upon reacting with a soluble tetrazolium dye. The detection can be continuous and suitable for kinetic studies. The kit provides a simple, straightforward protocol for a complete assay. Alternative name: Histone acetyltransferase. Applications: Colorimetric detection Activity assay.
Catalog No. 50-201-0162
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.