Learn More
Creative Biomart Recombinant Human HAVCR1 Antigen, His-tagged
Supplier:  Creative Biomart HAV090
Encompass_Preferred
HAVCR1 Human Recombinant produced in E. coli is a single polypeptide chain containing 298 amino acids (21-295 aa) and having a molecular mass of 31.9 kDa.HAVCR1 is fused to a 23 amino acid His-tag at
Catalog No. 50-208-4468
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.