Learn More
Creative Biomart Recombinant Human HAVCR2 Antigen, hIgG/His-tagged
Supplier:  Creative Biomart HAV094
Encompass_Preferred
Recombinant Human HAVCR2 produced in Sf9 Baculovirus cells is a single, glycosylated polypeptide chain containing 423 amino acids (aa 22-202) and having a molecular mass of 47.3 kDa.HAVCR2 is fused to
Catalog No. 50-208-4472
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.