Description
Haws drench showers feature the AXION MSR hydrodynamic shower design that minimizes the strong physical pressure that drives water to the outer rim of normal shower heads, making the distribution equal across the entire footprint of the flow. Better use of the water flowing through the shower makes it more effective while providing a more comfortable environment by breaking the streams of water into smaller individual droplets.
Specifications
Specifications
| Certifications/Compliance | ANSI Z358.1, CSA |
| Flow Rate | 20 gpm |
| For Use With (Application) | Quick Drenching or Cleansing of the Body |
| Includes | AXION MSR™ hydrodynamic design Stainless Steel, Chrome Plated Brass stay open ball valve, Stainless Steel pull rod with triangular handle, universal sign |
| Mounting | Horizontal or Vertical |
| Product Type | Drench Shower |