Learn More
Haws™ Filter
SDP
Catalog No. 18888260 Shop All Haws Corporation Products
Change view
Click to view available options
Haws™ Filter
Catalog No.
18-888-260 Haws™ Filter
1 options
Catalog No. 18-888-260 Supplier Haws™ Supplier No. 9070
Add to Cart
Add to Cart
Haws™ Filter is for installation on all eyewash units for additional protection for eyes.
Model 9070 Filter is recommended for installation on all eyewash units for additional protection for eyes. Made of FDA approved polypropylene with disposable FDA approved viscose fiber filter media which removes particles down to 20 microns. Flow Capacity: 7 gpm (26.5 L) max (recommended). Maximum Working Pressure: 125 psi (8.62 bar) Size: 15 in. (38.1 cm) overall by 5 in. (12.7 cm) diameter Supply: 3/4 in. NPT inlet and outlet.
Warranty and Services
1 year from ship date (Limited)
Specifications
Color Blue, Black
Height (English) 12.0625 in.
Height (Metric) 30.5 cm
Product Type Filter