Learn More
Haws™ In-Line Strainer
SDP
Catalog No. 18888278 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Chrome-plated Brass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-888-278 Chrome-plated Brass
18-888-279 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-888-278 Supplier Haws™ Supplier No. SP509
Add to Cart
Add to Cart
Haws™ In-Line Strainer is a 50x50 mesh strainer that filters debris from reaching the eye or eye/face wash
Model SP509 is a fully serviceable in-line, 50 x 50 mesh strainer filters debris from reaching the eye or eye/face wash.
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) 7260B-7270B, 7260BT-7270BT, 7261-7271, 7360B-7460B, 7360BT-7460BT, 7360BTWC, 7361-7461, 7460, 8300-8309, 8300CRP-8309CRP, 8320-8325, 8320CRP, 8330, Eye and Face Wash
Material Chrome-plated Brass
Product Type In-line Strainer
Weight (English) 1 lb.
Weight (Metric) 0.453592 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov