Learn More
National Marker™ Hazard Communications Material Safety Data Sheet Poster
Shop All National Marker Company ProductsDescription
This colorful, wall-size poster shows employees how to use and understand Material Safety Data Sheets, helping employers meet the requirements of the OSHA Hazard Communication Standard. Includes bright illustrations and brief explanations of the sections frequently included on an MSDS.
Specifications
Specifications
| Format | Poster |
| Product Type | MSDS Poster - English |
| For Use With (Application) | Provides detailed information about regulations and best practices |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.