Learn More
Fisherbrand™ Hazardous Chemical Dispensers
SCI_ED
Catalog No. S29124 Shop All Fisherbrand Products
Change view
Click to view available options
Description:
Neck size: 45 mm
Catalog No. Description
S29124 Neck size: 45 mm
1 options
This item is not returnable. View return policy
Catalog No. S29124
This item is not returnable. View return policy
Bottle-top dispensers for use with hazardous and corrosive chemicals.
  • Constructed entirely of PTFE with totally inert pouring surfaces—even the neck-sealing ring is encapsulated in PTFE
  • Incorporated breather vent allows for direct and controlled pourings
  • Captured PTFE closure ensures contaminant-free contents when not in use
  • Fit standard reagent bottles with neck sizes of GL32, 38, and 45
Specifications
Description Neck size: 45 mm