Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Communication & Right-to-know Labels - Coated Paper

Catalog No. 19110431 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
0.875 x 2.875 in.
1 x 3 in.
3.875 x 1.5 in.
4 x 5.875 in.
6 x 6 in.
7 x 10 in.
7 x 7 in.
7 x 8 in.
Display Text:
CHEMICAL NAME COMMON NAME MANUFACTURER DATE
CHEMICAL NAME: HEALTH HAZARD FIRE HAZARD SPECIFIC HAZARD INSTABILITY
CHEMICAL: OWNER: DATE:
HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT
IDENTITY: CAS NO.: HEALTH FLAMMABILITY INSTABILITY PROTECTIVE EQUIPMENT
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Display Text
19-110-431 7 x 8 in. CHEMICAL NAME: HEALTH HAZARD FIRE HAZARD SPECIFIC HAZARD INSTABILITY
19-110-380 0.875 x 2.875 in. CHEMICAL: OWNER: DATE:
19-110-329 1 x 3 in. CHEMICAL NAME COMMON NAME MANUFACTURER DATE
19-110-425 7 x 10 in. HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT
19-110-424 6 x 6 in. HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT
19-110-427 3.875 x 1.5 in. CHEMICAL NAME COMMON NAME MANUFACTURER DATE
19-110-430 7 x 7 in. IDENTITY: CAS NO.: HEALTH FLAMMABILITY INSTABILITY PROTECTIVE EQUIPMENT
19-110-428 4 x 5.875 in. HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-110-431 Supplier Brady™ Supplier No. 99425
Add to Cart
Add to Cart

Coated Paper (B-235) features surface printed graphics on semi gloss paper label stock making it ideal for general label applications.

  • B-235 Coated Paper with pressure sensitive acrylic adhesive from Brady™ adheres to most surfaces.
  • Labels remove cleanly, leaving no residue on surface.
  • Operating temperature of -40°F to +150°F (-40°C to +65.5°C).
  • Recommended for indoor use.

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS), NFPA 704, OSHA 1910.1200
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Color Black, Blue, Red, Yellow on White
Finish Semi-gloss
Format Label
Material Paper
Printable Sides Single-side Printable
Product Type Label
Sterility Non-sterile
Display Text CHEMICAL NAME: HEALTH HAZARD FIRE HAZARD SPECIFIC HAZARD INSTABILITY
Includes Pack of 100 Labels
Languages English
No. per Pack 100 Labels
Quantity 100/Roll
Shape Round
Size (English) 8 x 7 in.
Thickness (English) 3.8 mil
Thickness (Metric) 0.097 mm
Dimensions (L x W) 7 x 8 in.
Width (English) 8 in.
Width (Metric) 203.20 mm
Height (English) 7 in.
Height (Metric) 177.80 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.