Learn More
Description
- Provides workers with chemical information of hazardous materials
- Made from B-928 vinyl material
- Label Size: W x H: 4.5 x 3.75 in.
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent, Cold-temperature |
| Color | Black, Yellow |
| Material | Vinyl (B-928) |
| Product Type | Hazardous Materials Label |
| Display Text | PROPIONALDEHYDE |
| Quantity | 25/Pk. |
| Shape | Rectangle |
| Size (English) | 4.5 x 3.75 in. (W x H) |
| Thickness (English) | 0.0049 in. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.