Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Shipping Labels: Black on White, CAUTION: UNSCREW THIS BUNG SLOWLY DO NOT UNSCREW ENTIRELY UNTIL…

Catalog No. 19107329 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
3 x 5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-107-329 3 x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-107-329 Supplier Brady™ Supplier No. 63243

Black on White, 3 H x 5 in W

Made of plastic

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Description Label
Adhesive Type Self-adhesive
Color Black on White
Format Label
Material Plastic
Printable Sides Single-side Printable
Product Type Label
Languages English
Quantity 1 Ea.
Shape Diamond
Size (English) 5 x 3 in. (W x H)
Dimensions (L x W) 3 x 5 in.
Width (English) 5 in.
Height (English) 3 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.