Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hazardous Materials Classification Reference
SCI_ED

Catalog No. S47352C Shop All AccuformNMC Products
Change view
Click to view available options
Width (English):
3.5 in.
10 in.
Material:
Plastic
Vinyl
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Material
S47352C 3.5 in. Plastic
S47352D 10 in. Vinyl
S47352A 10 in. Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S47352C Supplier Accuform Signs Supplier No. HMC3R

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Hazmat chart is perfect for training or quick reference.

Graphically explains the NFPA hazard identification coding system.

Specifications

Material Plastic
For Use With (Application) For training or quick reference, graphically explains the NFPA hazard identification coding system
Height (English) 2.5 in.
Height (Metric) 6.4 cm
Width (English) 3.5 in.
Width (Metric) 8.9 cm
Product Type Chemical Safety Chart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.