Learn More
CTL Scientific Supply Corp HAZMAT 3 ITEM PACK
SDP
Supplier:  CTL Scientific Supply Corp HM100
Encompass_Preferred
Pack containing: 1 box F-100 Fluoride Test Paper, 200 strips (20 x 70 mm), 1 box 90758 Potassium Iodide Starch Paper, 200 strips (20 x 70 mm), 1 box UNIV-114LD Universal 1-14 pH paper, roll (26' x 5/8 in.).
Catalog No. NC9072042