Learn More
New Pig HazMat Chemical Absorbent Mat Pad in Dispenser Box
SDP
Catalog No. 18389135 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: HazMat Chemical Absorbent Mat Pad in Dispenser Box
Catalog No.
18-389-135 Type: HazMat Chemical Absorbent Mat Pad in Dispenser Box
1 options
This item is not returnable. View return policy
Catalog No. 18-389-135 Supplier New Pig Supplier No. MAT354
  • $192.00
    $152.65 / Case of 100
    Save $39.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent pads; dispenser box is easy to store.
  • Clean up big spills or persistent leaks of hazardous chemicals with our most popular mat weight; pads come in an easy-to-store dispenser box
  • Soaks up the widest range of acids, bases, corrosives, chemicals and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Absorbs a wide range of hazardous liquids found in chemical storage areas, around dip tanks, in cleaning agents used in food processing plants, in boiler rooms, and labs
  • Pink color is designated for hazardous chemicals and easy to identify in a spill emergency or cleanup
  • Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • PIG Mat lasts 2x longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces