Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig HazMat Chemical Absorbent Mat Roll
SDP

Catalog No. 18813354 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
20.1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-813-354 20.1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-813-354 Supplier New Pig Supplier No. MAT303
  • $224.00
    $199.65 / Each of 1

    Save $24.35

    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heavy weight, 15 in. x 150 ft. (W x L), 1 roll per bag.

  • Safely soak up acids, bases, corrosives and hazardous chemical spills
  • Soaks up a wide range of acids, bases, corrosives, chemicals and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide

Specifications

Capacity (English) 20.1 gal.
Capacity (Metric) 76.1 L
Material Polypropylene
Type Hazmat Absorbent Pad
Disposable Single-use
Weight Capacity Heavyweight
Length (English) 16 in.
Length (Metric) 41 cm
Width (English) 16 in.
Width (Metric) 41 cm
Weight (English) 16 lb.
Weight (Metric) 7.3 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.