Learn More
Description
- Mats are perforated at the bound edge and center for versatile sizing
- Pop-up tabs on cardboard back let you hang pack
- Safely soak up acids, bases, corrosives and hazardous chemical spills
- Pink color is designated for hazardous chemicals and easy to identify in a spill emergency or cleanup
- Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups
- Multiple layers of thermally bonded 100% polypropylene
- Dimple pattern speeds wicking of liquid throughout mat
- Dimensions: 14 x 15 in.
- 9 tablets with 10 pads each for a total of 90 pads
Specifications
Specifications
| Absorbancy | 13.86 gal. |
| Color | Pink |
| Material | Polypropylene |
| Type | Chemical Absorbent Pad |
| Disposable | Single-use |
| Packaging Quantity | 90/Cs. |
| Weight Capacity | Heavyweight |
| Length (English) | 15 in. |
| Length (Metric) | 38 cm |
| Width (English) | 14 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
